Anti-DDX53 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
034559-AP
USBI034559-APEA 884 EUR
USBI034559-AP
Anti-DDX53 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: DDX53 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 53)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR