Anti-DUS4L Rabbit Polyclonal Antibody

Lieferant: US Biological
034850
USBI034850EA 649 EUR
USBI034850
Anti-DUS4L Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: DUS4L (dihydrouridine synthase 4-like (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR