Anti-EOMES Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
035100-AP
USBI035100-APEA 884 EUR
USBI035100-AP
Anti-EOMES Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: EOMES (eomesodermin)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human