Anti-EOMES Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
035100-APC
USBI035100-APCEA 884 EUR
USBI035100-APC
Anti-EOMES Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: EOMES (eomesodermin)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human