Anti-SRSF10 Rabbit Polyclonal Antibody

Lieferant: US Biological
035795
USBI035795EA 667 EUR
USBI035795
Anti-SRSF10 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SRSF10 (serine/arginine-rich splicing factor 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR