Anti-GCFC Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
035956-BIOTIN
USBI035956-BIOTINEA 884 EUR
USBI035956-BIOTIN
Anti-GCFC Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: GCFC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR