Anti-GNAZ Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
036136-APC
USBI036136-APCEA 884 EUR
USBI036136-APC
Anti-GNAZ Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: GNAZ (guanine nucleotide binding protein (G protein), alpha z polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human