Anti-GNE Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: US Biological
036141-HRP
USBI036141-HRPEA 884 EUR
USBI036141-HRP
Anti-GNE Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: GNE (glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR