Anti-GNE Rabbit Polyclonal Antibody

Lieferant: US Biological
036142
USBI036142EA 649 EUR
USBI036142
Anti-GNE Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: GNE (glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR