Anti-HEMGN Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
036505-BIOTIN
USBI036505-BIOTINEA 884 EUR
USBI036505-BIOTIN
Anti-HEMGN Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: HEMGN (hemogen)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human