Anti-HERPUD1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
036516-AP
USBI036516-APEA 884 EUR
USBI036516-AP
Anti-HERPUD1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HERPUD1 (homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human