Anti-HERPUD1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
036516-APC
USBI036516-APCEA 884 EUR
USBI036516-APC
Anti-HERPUD1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HERPUD1 (homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human