Anti-HEYL Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
036527-APC
USBI036527-APCEA 884 EUR
USBI036527-APC
Anti-HEYL Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HEYL (hes-related family bHLH transcription factor with YRPW motif-like)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human