Anti-HOXD12 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
036730-BIOTIN
USBI036730-BIOTINEA 884 EUR
USBI036730-BIOTIN
Anti-HOXD12 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: HOXD12 (homeobox D12)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human