Anti-HOXD12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: US Biological
036730-FITC
USBI036730-FITCEA 884 EUR
USBI036730-FITC
Anti-HOXD12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: HOXD12 (homeobox D12)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR