Anti-HRC Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
036759-AP
USBI036759-APEA 884 EUR
USBI036759-AP
Anti-HRC Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: HRC (histidine rich calcium binding protein)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR