Anti-HSD11B2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
036776-BIOTIN
USBI036776-BIOTINEA 884 EUR
USBI036776-BIOTIN
Anti-HSD11B2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: HSD11B2 (hydroxysteroid (11-beta) dehydrogenase 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR