Anti-HSPB9 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
036808-PE
USBI036808-PEEA 884 EUR
USBI036808-PE
Anti-HSPB9 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HSPB9 (heat shock protein, alpha-crystallin-related, B9)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human