Anti-IP6K3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
037070-PE
USBI037070-PEEA 884 EUR
USBI037070-PE
Anti-IP6K3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: IP6K3 (inositol hexakisphosphate kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human