Anti-IRX5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
037115-PE
USBI037115-PEEA 884 EUR
USBI037115-PE
Anti-IRX5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: IRX5 (iroquois homeobox 5)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR