Anti-KDM3B Rabbit Polyclonal Antibody

Lieferant: US Biological
037195
USBI037195EA 649 EUR
USBI037195
Anti-KDM3B Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Dot blot 1:100-500

Type: Primary
Antigen: KDM3B (lysine (K)-specific demethylase 3B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR