Anti-KIF3B Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
037442-BIOTIN
USBI037442-BIOTINEA 884 EUR
USBI037442-BIOTIN
Anti-KIF3B Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: KIF3B (kinesin family member 3B)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR