Anti-LCE1A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
037708-AP
USBI037708-APEA 884 EUR
USBI037708-AP
Anti-LCE1A Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: LCE1A (late cornified envelope 1A)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human