Anti-LCE1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
037708-APC
USBI037708-APCEA 884 EUR
USBI037708-APC
Anti-LCE1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: LCE1A (late cornified envelope 1A)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR