Anti-LCE1A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
037708-PE
USBI037708-PEEA 884 EUR
USBI037708-PE
Anti-LCE1A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: LCE1A (late cornified envelope 1A)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human