Anti-MAP1ALC3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
038090-BIOTIN
USBI038090-BIOTINEA 884 EUR
USBI038090-BIOTIN
Anti-MAP1ALC3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100

Type: Primary
Antigen: MAP1ALC3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR