Anti-MAP1LC3C Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
038116-BIOTIN
USBI038116-BIOTINEA 884 EUR
USBI038116-BIOTIN
Anti-MAP1LC3C Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100
Type: Primary
Antigen: MAP1LC3C (microtubule-associated protein 1 light chain 3 gamma)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human