Anti-MARCO Rabbit Polyclonal Antibody

Lieferant: US Biological
038200
USBI038200EA 487 EUR
USBI038200
Anti-MARCO Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR