Anti-MARCO Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
038200-PE
USBI038200-PEEA 616 EUR
USBI038200-PE
Anti-MARCO Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR