Anti-SERPINB5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
038214-PE
USBI038214-PEEA 884 EUR
USBI038214-PE
Anti-SERPINB5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Do not freeze!

Type: Primary
Antigen: SERPINB5 (serpin peptidase inhibitor, clade B (ovalbumin), member 5)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR