Anti-MBD4 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
038241-BIOTIN
USBI038241-BIOTINEA 884 EUR
USBI038241-BIOTIN
Anti-MBD4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MBD4 (methyl-CpG binding domain protein 4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human