Anti-MCTP2 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
038293-BIOTIN
USBI038293-BIOTINEA 884 EUR
USBI038293-BIOTIN
Anti-MCTP2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MCTP2 (multiple C2 domains, transmembrane 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human