Anti-OR2L8 Rabbit Polyclonal Antibody

Lieferant: US Biological
039361
USBI039361EA 649 EUR
USBI039361
Anti-OR2L8 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: OR2L8 (olfactory receptor, family 2, subfamily L, member 8 (gene/pseudogene))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR