Anti-OR5L2 Rabbit Polyclonal Antibody

Lieferant: US Biological
039459
USBI039459EA 649 EUR
USBI039459
Anti-OR5L2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: OR5L2 (olfactory receptor, family 5, subfamily L, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR