Anti-PAK7 Rabbit Polyclonal Antibody

Lieferant: US Biological
039629
USBI039629EA 667 EUR
USBI039629
Anti-PAK7 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: PAK7 (p21 protein (Cdc42/Rac)-activated kinase 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR