Anti-PEG10 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
039870-AP
USBI039870-APEA 884 EUR
USBI039870-AP
Anti-PEG10 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: PEG10 (paternally expressed 10)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR