Anti-PEG10 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
039870-APC
USBI039870-APCEA 884 EUR
USBI039870-APC
Anti-PEG10 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: PEG10 (paternally expressed 10)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human