Anti-PI3KC3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
039981-BIOTIN
USBI039981-BIOTINEA 884 EUR
USBI039981-BIOTIN
Anti-PI3KC3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PI3KC3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR