Anti-PIK3R5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
040057-AP
USBI040057-APEA 884 EUR
USBI040057-AP
Anti-PIK3R5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: PIK3R5 (phosphoinositide-3-kinase, regulatory subunit 5)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR