Anti-PIK3R5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: US Biological
040057-FITC
USBI040057-FITCEA 884 EUR
USBI040057-FITC
Anti-PIK3R5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: PIK3R5 (phosphoinositide-3-kinase, regulatory subunit 5)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human