Anti-PINK1 Rabbit Polyclonal Antibody

Lieferant: US Biological
040070
USBI040070EA 649 EUR
USBI040070
Anti-PINK1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot1:100-500

Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR