Anti-PINK1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
040072-AP
USBI040072-APEA 884 EUR
USBI040072-AP
Anti-PINK1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse