Anti-NRIP1 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
041059-BIOTIN
USBI041059-BIOTINEA 884 EUR
USBI041059-BIOTIN
Anti-NRIP1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: NRIP1 (nuclear receptor interacting protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human