Anti-RNH1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
041156-PE
USBI041156-PEEA 884 EUR
USBI041156-PE
Anti-RNH1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50
Do not freeze!

Type: Primary
Antigen: RNH1 (ribonuclease/angiogenin inhibitor 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR