Anti-RWDD3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: US Biological
041342-FITC
USBI041342-FITCEA 884 EUR
USBI041342-FITC
Anti-RWDD3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: RWDD3 (RWD domain containing 3)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human