Anti-SPR Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
042258-PE
USBI042258-PEEA 884 EUR
USBI042258-PE
Anti-SPR Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SPR (sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR