Anti-STK4 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
042413-BIOTIN
USBI042413-BIOTINEA 884 EUR
USBI042413-BIOTIN
Anti-STK4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: STK4 (serine/threonine kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse