Anti-STS Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
042426-BIOTIN
USBI042426-BIOTINEA 884 EUR
USBI042426-BIOTIN
Anti-STS Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: STS (steroid sulfatase (microsomal), isozyme S)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR