Anti-TAS2R38 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
042614-APC
USBI042614-APCEA 884 EUR
USBI042614-APC
Anti-TAS2R38 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TAS2R38 (taste receptor, type 2, member 38)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR