Anti-TEC Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
042727-PE
USBI042727-PEEA 884 EUR
USBI042727-PE
Anti-TEC Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: TEC (tec protein tyrosine kinase)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human